Learn More
Invitrogen™ Candida albicans Polyclonal Antibody, Biotin

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173156
Description
The PA1-73156 antibody reacts with Candida albicans. PA1-73156 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73156 antibody was raised against Candida albicans, type A.
Candida albicans is the most frequently isolated fungal pathogen of humans, affecting immuocompromised patients ranging from premature infants to AIDS sufferers. Systemic infections have an attributed mortality of 30-50%. C. albicans is a diploid organism which has eight sets of homologous chromosomes. Its genome size is about 16 Mb (haploid), about 30% greater than S. cerevisiae (baker's yeast).
Specifications
| Candida albicans | |
| Polyclonal | |
| Biotin | |
| C. albicans | |
| Candida albicans, type A. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Yeast | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.