Learn More
Novus Biologicals™ Canine Universal Protein Whole Tissue Lysate (Adult Whole Normal)
Product Code. 18260022 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
4 x 0.5 mg
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18260022 4 x 0.5 mg Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18260022 Supplier Novus Biologicals™ Supplier No. NB82089205
The Canine Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from canine. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Western Blot
Tissue Universal
Quantity 4 x 0.5 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Universal Protein Lysate: Dog Normal Tissues
Cellular Fraction Whole
Tissue Condition Normal
Cross Reactivity Dog
Host Species Canine
Concentration 5 mg/mL
Product Type Canine Universal Protein Whole Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native