Learn More
Paragon Scientific™ Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids (UKAS ISO 17025 certified)
Product Code. 15929979
Change view
Click to view available options
Viscosity Range:
0.4 to 2 cSt
0.8 to 4 cSt
1.6 to 8 cSt
10,000 to 50,000 cSt
100 to 500 cSt
1600 to 8000 cSt
20 to 100 cSt
20,000 to 100,000 cSt
240 to 1200 cSt
3 to 15 cSt
4000 to 20,000 cSt
50 to 100 cSt
500 to 2500 cSt
7 to 35 cSt
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range Unit Size
15929979 100 to 500 cSt Each
15989959 0.4 to 2 cSt Each
15909969 0.8 to 4 cSt Each
15929969 1.6 to 8 cSt Each
15949969 3 to 15 cSt Each
15969969 7 to 35 cSt Each
15989969 20 to 100 cSt Each
15909979 50 to 100 cSt Each
15949979 240 to 1200 cSt Each
15969979 500 to 2500 cSt Each
15989979 1600 to 8000 cSt Each
15909989 4000 to 20,000 cSt Each
15929989 10,000 to 50,000 cSt Each
15949989 20,000 to 100,000 cSt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15929979 Supplier Paragon Scientific™ Supplier No. CFO08UKC
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For the determination of kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
  • Supplied with full UKAS ISO 17025 calibration
Specifications
Viscosity Range 100 to 500 cSt
Calibrated Yes
Approximate Constant 0.5
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D445, ISO 3104 and IP 71 Section 1
Includes UKAS ISO 17025 calibration
Size 350
8 week lead-time