Learn More
Applied Biosystems™ Capillary Cutter, 310 Genetic Analyzer
Product Code. 17142959 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2 Each
Unit Size:
Pack of 2
Product Code. Quantity unitSize
17142959 2 Each Pack of 2
1 options
Product Code. 17142959
Brand: Applied Biosystems™ 401958
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This is a capillary cutter for use with 310 genetic analyzer capillaries. Shortened capillaries can be used for short run modules.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Product Type Capillary Cutter
Quantity 2 Each
Shipping Condition Room Temperature