Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Product Code. 10161721
Change view
Click to view available options
Length (Metric) Tube:
60 mm
70 mm
75 mm
80 mm
90 mm
100 mm
120 mm
150 mm
Diameter (Metric) Inner:
0.80 mm
0.95 mm
1.15 mm
1.30 mm
1.55 mm
Unit Size:
Pack of 1000
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tube Diameter (Metric) Inner Unit Size
10161721 70 mm 1.55 mm Pack of 1000
10769191 70 mm 0.80 mm Pack of 1000
10673192 75 mm 0.80 mm Pack of 1000
10033961 80 mm 0.80 mm Pack of 1000
10355511 90 mm 0.80 mm Pack of 1000
10253861 120 mm 0.80 mm Pack of 1000
10720233 60 mm 0.95 mm Pack of 1000
10223531 90 mm 0.95 mm Pack of 1000
10006854 100 mm 0.95 mm Pack of 1000
10380182 120 mm 0.95 mm Pack of 1000
10009760 70 mm 1.15 mm Pack of 1000
10613202 120 mm 1.15 mm Pack of 1000
10029770 150 mm 1.15 mm Pack of 1000
10089650 70 mm 1.30 mm Pack of 1000
10673002 75 mm 1.30 mm Pack of 1000
10009750 100 mm 1.30 mm Pack of 1000
10223341 75 mm 1.55 mm Pack of 1000
10409712 80 mm 1.55 mm Pack of 1000
10089730 90 mm 1.55 mm Pack of 1000
10233671 100 mm 1.55 mm Pack of 1000
10059720 120 mm 1.55 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Product Code. 10161721 Supplier Hirschmann™ Supplier No. 9212370
  • ONLINE EXCLUSIVE PRICE
    142.00€
    123.00€ / Pack of 1000
    Save 19.00€
    13% Off
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Excellent for melting point analysis in the laboratory
  • Capillary tubes
  • One end closed
  • Available in a wide range of diameters/lengths
Specifications
Product Type Melting Point Determination Tube, One End Closed
Diameter (Metric) Inner 1.55 mm
Length (Metric) Tube 70 mm
Quantity 1 x 1000/Pk.