Learn More
CAPN3 Mouse anti-Human, Unconjugated, Clone: 1G12A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67366-1-IG-150UL
Description
The CAPN3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CAPN3. This antibody recognizes human, pig, mouse, rat antigen. The CAPN3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CAPN3 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, ELISA | |
| 1G12A5 | |
| Unconjugated | |
| 150 μL | |
| 825 | |
| IgG1 |