missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPNS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25057-1-AP-150UL
This item is not returnable.
View return policy
Description
Calpains are a ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. Calpain families have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. Calpain I and II are heterodimeric with distinct large subunits associated with common small subunits, all of which are encoded by different genes. Calpain S1 is a small subunit common to both calpain I and II and is associated with myotonic dystrophy.Specifications
| CAPNS1 | |
| Polyclonal | |
| Unconjugated | |
| CAPNS1 | |
| 30K, Calpain regulatory subunit, Calpain small subunit 1, calpain, small subunit 1, CALPAIN4, CANP, CANP small subunit, CANPS, CAPN4, CAPNS, CAPNS1, CDPS, CSS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12336, 826 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88456, P04632 | |
| CAPNS1 | |
| CAPNS1 Fusion Protein Ag21885 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction