missing translation for 'onlineSavingsMsg'
Learn More

Caprylic Acid, Mikromol™

Product Code. 17912536
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17912536 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17912536 Supplier Mikromol Supplier No. MM3387.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Caprylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Caprylic Acid
CAS 124-07-2
Analyte or Component Names Caprylic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H16 O2
Certifications/Compliance ISO 17025
Synonym Octanoic acid, 1-Heptanecarboxylic acid, Caprylic acid, Edenor C 8-98-100, Emery 657, Kortacid 0899, Lunac 8-95, Lunac 8-98, NAA 82, NSC 5024, Neo-Fat 8, Neo-Fat 8S, Octylic acid, Prifac 2901, n-Caprylic acid, n-Octanoic acid, n-Octoic acid, n-Octylic acid, 8:0 caprylic acid
InChI Formula InChI=1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)
SMILES CCCCCCCC(=O)O
IUPAC Name octanoic acid
Molecular Weight (g/mol) 144.21
Quantity 250 mg
Formula Weight 144.115 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.