Learn More
Erlab™ Captair 822B Smart Filtering Storage Cabinet
Erlab™ Captair 822B Smart Filtering Storage Cabinets provide a steady flow of air through the cabinet, capturing hazardous vapors in the filters, and returns clean air back to the room.
3700.00€ - 3835.00€
Specifications
Certifications/Compliance | Filtration performances tested according to the AFNOR NF X 15-211:2009 standard : France EN 1822 : 1998 (HEPA H14 & ULPA U17 Filters) - CE Marking |
---|---|
Description | Air Flow: 70 m3/h |
Dimensions (D x W x H) | 580 x 650 x 755 mm |
Product Type | Ductless Filtering Storage Cabinet |
Holds | 20 to 25 bottles |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
15544974
|
Erlab™
B03IA02B/AS |
Filter AS |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15564974
|
Erlab™
B03IA02B/F |
Filter F |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Filter AS - For organic vapors
- Filter BE - For acid vapors
- Filter F - For formaldehyde vapors
- Filter K - For ammonia vapors
- HP - HEPA H14:99.995 % efficiency filtration of particles over 0.1 μm in size
- 80 to 220 V, 50/60 Hz, 35 W
- Corrosion resistant electro-galvanized steel coated with anti-acid polymere
- Doors: Clear, chemical resistant acrylic for easy viewing
- Filtration Module: Polypropylene
- 2 storage compartments with sliding doors or 1 storage compartment with large volume drawer
- Simple communication by LED pulsation system: fan settings, usage timer, fan failure, face velocity, automatic filter saturation detection
- App for remote control to monitor the chemical storage cabinet, change the settings, and deliver safety salerts immediately to your devices (mobile, tablet and PC)
- RJ45 cable connection
- Detection sensors for solvents or acids or formaldehyde
- Alarm if doors are left open
Specifications
Filtration performances tested according to the AFNOR NF X 15-211:2009 standard : France EN 1822 : 1998 (HEPA H14 & ULPA U17 Filters) - CE Marking | |
580 x 650 x 755 mm | |
20 to 25 bottles |
Air Flow: 70 m3/h | |
Ductless Filtering Storage Cabinet | |
Up to 25 x 1 L Bottles |