missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ Blue Chromatography Media

Product Code. 11797444 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11797444 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11797444 Supplier Cytiva Supplier No. 17544801
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

An affinity medium for capture of proteins and enzyme purification on large and small scale

This product is chemically stable for cleaning with strong agarose base matrix.

  • Chemical stability ensures tolerance of harsh solvents used in repeated CIP and sanitization procedures
  • Rigid agarose base matrix allows high flow rates and processing of large sample volumes with no reduction in binding capacity
  • Can be autoclaved repeatedly
  • Average particle size of 75μm

For purification of enzymes, capture of albumin and recombinant proteins at laboratory and process scales

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
Stability Stable in commonly used aqueous Buffers: 0.5 M NaOH, 8 M Urea, 6 M Guanidine Hydrochloride
For Use With (Application) For Enzyme PurifiCatIon, Capture of Albumin
Storage Requirements 2°C to 8°C
Binding Capacity >24 mg HSA/mL mediμm
Product Type Capto Blue Media
Flow Rate 600 cm/Hoμr
Particle Size 75 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Includes Certificate of Analysis
Max. Pressure 300 kPa
Matrix Highly Cross-Linked Agarose
pH Range 2 to 13
Storage Buffer 20% Ethanol
Ligand Type Cibacron Blue
Show More Show Less
compliance-icons
Product Identifier
  • 25 ML CAPTO BLUE
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.