Learn More
CAPZA1 Mouse anti-Canine, Human, Rat, Clone: OTI7C8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501125
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.Specifications
| CAPZA1 | |
| Monoclonal | |
| Unconjugated | |
| CAPZA1 | |
| CAPPA1, CAPZ, CAZ1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 475857, 691149, 829 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI7C8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| B2GUZ5, P52907 | |
| CAPZA1 | |
| Full length human recombit protein of human CAPZA1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.