Learn More
Invitrogen™ CAR Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558537
 View more versions of this product
Product Code. 30284580
  • 477.00€ / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Med. Bio. Labs patent anti-CXADR.
The protein encoded by this gene is a type I membrane receptor for group B coxsackieviruses and subgroup C adenoviruses. Pseudogenes of this gene are found on chromosomes 15, 18, and 21.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CAR Biosimilar
Recombinant Monoclonal
Unconjugated
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG2SA