missing translation for 'onlineSavingsMsg'
Learn More

Carbendazim 50 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30323702
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323702 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323702 Supplier Dr. Ehrenstorfer Supplier No. DREA10990000ME50
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbendazim
CAS 10605-21-7
Analyte or Component Names Carbendazim
Shipping Condition +20°C
Molecular Formula C9 H9 N3 O2
Concentration 50 μg/mL
Certifications/Compliance ISO 17034
Synonym Albendazole Imp. E (EP), Fenbendazole Imp. A (EP), Fenbendazole USP Related Compound A, Fenbendazole USP RC A, Carbendazim, Methyl (1H-Benzimidazol-2-yl)carbamate, 1H-Benzimidazol-2-ylcarbamic acid methyl ester, 2-Benzimidazolecarbamic acid methyl ester, 2-(Carbomethoxyamino)benzimidazole, 2-(Methoxycarbamoyl)benzimidazole, 2-(Methoxycarboxamido)benzimidazole, 2-[(Methoxycarbonyl)amino]benzimidazole, A 118, A 118 (pesticide), Agrizim, Antibac MF, BAS 3460, BAS 3460F, BAS 67054F, BCM, BCM (fungicide), BMK, BMK (fungicide), Battal, Bavistan, Bavistin, Bavistin 25SD, Bavistin 3460, Bavistin 50SD, Bavistin FL, Bavistine, Bengard, Bercema-Bitosen, Bitosen, CTR 6669, Carben VL, Carbendazime, Carbendazol, Carbendazole, Delsene, Delsene 10, Derosal, Derosal 60PM, EK 578, Falicarben, Funaben, Funaben 3, Funaben 50, Fungoxan, G 665, Garbenda, HOE 17411, IPO 1250, IPO Y, Jkstein, Karben, Karben Stefes Flo, Karben flo Stefes, Kolfugo, Kolfugo 25FW, Kolfugo Extra, Lignasan, MBC, Mecarzole, Medamine, Methoxycarbonylaminobenzimidazole, Methyl 1H-benzimidazol-2-ylcarbamate, Methyl 1H-benzimidazole-2-carbamate, Methyl 2-benzimidazolecarbamate, Methyl 2-benzimidazolylcarbamate, Methyl benzimidazolylcarbamate, Myco, NSC 109874, Olgin, Olgin (fungicide), Preparation G 665, Preventol BCM, Protek, Sarfun, Spin, Spin (pesticide), Stein, Subeej, Supercarb, Thicoper, Triticol, U 32104, Zhiweiling, SKF-26058, Ph Eur Albendazole Impurity E, Fenbendazole Related Compound A, Albendazole Impurity E, Fenbendazole Impurity A
InChI Formula InChI=1S/C9H9N3O2/c1-14-9(13)12-8-10-6-4-2-3-5-7(6)11-8/h2-5H,1H3,(H2,10,11,12,13)
SMILES COC(=O)Nc1nc2ccccc2[nH]1
IUPAC Name methyl N-(1H-benzimidazol-2-yl)carbamate
Solvent or Matrix Methanol
Molecular Weight (g/mol) 191.19
Quantity 1 mL
Formula Weight 191.0695 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.