missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Carbenicillin ReadyMade™ Solution

Product Code. 30044026
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
50 x 1 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044026 10 x 1 mL 10mL
30044041 50 x 1 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30044026 Supplier TOKU-E™ Supplier No. C28910x1ML
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Carbenicillin ReadyMade™ Solution is provided as a filter-sterilized solution of Carbenicillin Disodium in 20-30% ethanol.

  • CAS Number: 4800-94-6
  • Molecular Formula: C17H16N2Na2O6S
  • Molecular Weight: 422.36 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Semi-synthetic
  • Spectrum: The activity of Carbenicillin is limited to primarily Gram-negative bacteria including Pseudomonas aeruginosa and common enteric bacteria. It has limited activity against Gram-positive bacteria
  • Mechanism of Action: Carbenicillin interferes with penicillin binding protein (PBP) activity otherwise involved in the final phase of peptidoglycan synthesis. PBPs are enzymes which catalyze a pentaglycine crosslink between alanine and lysine residues. Without a pentaglycine crosslink, the integrity of the cell wall is severely compromised ultimately leading to cell lysis. Resistance to β-lactams is commonly due to cells containing plasmid-encoded β-lactamases

Specifications

Chemical Name or Material Carbenicillin
Physical Form Liquid
CAS 4800-94-6
Source Semi-synthetic
Packaging 2 mL plastic vial
Sterile Filtered Yes, 0.22 μm
Sterility Sterile
Color Colorless
Concentration 100 to 105 mg/mL
Quantity 10 x 1 mL
Flash Point 37.8°C (104°F)-closed cup
Purity Grade Notes >95% (HPLC)
Assay Percent Range >770 μg/mg
Assay Percent Notes Carbenicillin DiNa, Dried Basis
Molecular Formula C17H16N2Na2O6S
Product Line ReadyMade™ Solutions
Shipping Condition Ice Pack
Synonym α-Carboxybenzylpenicillindisodium salt; Pyopen; α-Carboxybenzylpenicillin
Solubility Information Soluble in Water
IUPAC Name disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 422.36 g/mol
Formula Weight 422.36 amu
Percent Purity >95%
Grade Antibiotic
Concentration or Composition (by Analyte or Components) Carbenicillin disodium salt ≥10 to <20%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.