missing translation for 'onlineSavingsMsg'
Learn More

Carbenoxolone Disodium, TRC

Product Code. 30368175
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368175 50 mg -
30361045 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30368175 Supplier Toronto Research Chemicals Supplier No. TRCC17588550MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7421-40-1
Molecular Formula C34 H48 O7 . 2 Na
Molecular Weight (g/mol) 614.72
Synonym Olean-12-en-29-oic acid, 3-(3-carboxy-1-oxopropoxy)-11-oxo-, sodium salt (1:2), (3β,20β)-, Olean-12-en-29-oic acid, 3-(3-carboxy-1-oxopropoxy)-11-oxo-, disodium salt, (3β,20β)- (9CI), Olean-12-en-30-oic acid, 3β-hydroxy-11-oxo-, hydrogen succinate, disodium salt (7CI,8CI), 3-(Succinyloxy)glycyrrhetic acid disodium salt, Bioral, Carbenoxalone disodium salt, Carbenoxalone sodium, Carbenoxolone disodium, Carbenoxolone disodium salt, Carbenoxolone sodium, Carbenoxolone sodium salt, Disodium glycyrrhetic acid hemisuccinate, Disodium glycyrrhetinyl succinate, Duogastrone, Glycyrrhetic acid hydrosuccinate disodium salt, Glycyrrhetinic acid hydrogen succinate disodium salt, Glycyrrhetinic acid succinate disodium salt, Neogel, Pyrogastrone, Sanodin, Sodium carbenoxolone, Ulcus-Tablinen
IUPAC Name disodium;(2S,4aS,6aR,6aS,6bR,8aR,10S,12aS,14bR)-10-(3-carboxylatopropanoyloxy)-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate
SMILES [Na+].[Na+].CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]3(C)[C@@H]2C(=O)C=C4[C@@H]5C[C@](C)(CC[C@]5(C)CC[C@@]34C)C(=O)[O-])OC(=O)CCC(=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C34H50O7.2Na/c1-29(2)23-10-13-34(7)27(32(23,5)12-11-24(29)41-26(38)9-8-25(36)37)22(35)18-20-21-19-31(4,28(39)40)15-14-30(21,3)16-17-33(20,34)6;;/h18,21,23-24,27H,8-17,19H2,1-7H3,(H,36,37)(H,39,40);;/q;2*+1/p-2/t21-,23-,24-,27+,30+,31-,32-,33+,34+;;/m0../s1
Formula Weight 614.3195
Chemical Name or Material Carbenoxolone Disodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.