missing translation for 'onlineSavingsMsg'
Learn More

Carbenoxolone, TRC

Product Code. 30138031
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138031 100 mg 100mg
30136948 500 mg 500mg
17938964 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138031 Supplier Toronto Research Chemicals Supplier No. TRCC175903100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5697-56-3
Molecular Formula C34 H50 O7
Molecular Weight (g/mol) 570.76
Synonym Olean-12-en-29-oic acid, 3-(3-carboxy-1-oxopropoxy)-11-oxo-, (3β,20β)-, Olean-12-en-30-oic acid, 3β-hydroxy-11-oxo-, hydrogen succinate (7CI,8CI), Olean-12-en-30-oic acid, 3β-hydroxy-11-oxo-, succinate (6CI), (3β,20β)-3-(3-Carboxy-1-oxopropoxy)-11-oxoolean-12-en-29-oic acid, 3-O-(β-Carboxypropionyl)-11-oxo-18β-olean-12-en-30-oic acid, 3β-Hydroxy-11-oxoolean-12-en-30-oic acid hydrogen succinate, Biogastrone, Carbenoxolone, Glycyrrhetinic acid hydrogen succinate, Glycyrrhetinic acid succinate
IUPAC Name (2S,4aS,6aR,6aS,6bR,8aR,10S,12aS,14bR)-10-(3-carboxypropanoyloxy)-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylic acid
SMILES CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]3(C)[C@@H]2C(=O)C=C4[C@@H]5C[C@](C)(CC[C@]5(C)CC[C@@]34C)C(=O)O)OC(=O)CCC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C34H50O7/c1-29(2)23-10-13-34(7)27(32(23,5)12-11-24(29)41-26(38)9-8-25(36)37)22(35)18-20-21-19-31(4,28(39)40)15-14-30(21,3)16-17-33(20,34)6/h18,21,23-24,27H,8-17,19H2,1-7H3,(H,36,37)(H,39,40)/t21-,23-,24-,27+,30+,31-,32-,33+,34+/m0/s1
Formula Weight 570.3557
Chemical Name or Material Carbenoxolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.