missing translation for 'onlineSavingsMsg'
Learn More

Carbimazole, TRC

Product Code. 30005521
Change view
Click to view available options
Quantity:
10 mg
1 g
Unit Size:
10mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005521 1 g 1g
30010612 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005521 Supplier Toronto Research Chemicals Supplier No. TRCC1759601G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22232-54-8
Molecular Formula C7 H10 N2 O2 S
Molecular Weight (g/mol) 186.23
Synonym Carbimazole, Ethyl 3-methyl-2-thioxo-2,3-dihydro-1H-imidazole-1-carboxylate, 1H-Imidazole-1-carboxylic acid, 2,3-dihydro-3-methyl-2-thioxo-, ethyl ester, 4-Imidazoline-1-carboxylic acid, 3-methyl-2-thioxo-, ethyl ester (6CI,8CI), 1-Ethoxycarbonyl-3-methyl-2-thio-4-imidazoline, 1-Methyl-3-carbethoxy-2-thioglyoxalone, 3-Carbethoxymercapto-1-methylimidazole, 3-Carbethoxythio-1-methylglyoxaline, 3-Carbethoxythio-1-methyliminazole, Athyromazole, Basolest, CG 1, Carbethoxymethimazole, Carbimazol, Carbimazole, Ethyl 3-methyl-2-thioimidazoline-1-carboxylate, Ethyl 3-methyl-2-thioxo-δ4-imidazoline-1-carboxylate, Neo-Thyreostat, Neomercazole, Tyrazol
IUPAC Name ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
SMILES CCOC(=O)N1C=CN(C)C1=S

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H10N2O2S/c1-3-11-7(10)9-5-4-8(2)6(9)12/h4-5H,3H2,1-2H3
Formula Weight 186.0463
Percent Purity >95%
Chemical Name or Material Carbimazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.