Learn More
Swann-Morton™ Carbon Steel Individual Foil Packs, Sterile Scalpel Blades, No. 9
Product Code. 15957682
Change view
Click to view available options
Quantity:
100/Box
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15957682 100/Box Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15957682 Supplier Swann-Morton™ Supplier No. 0217
  • ONLINE EXCLUSIVE PRICE
    21.55€
    18.84€ / Pack of 100
    Save 2.71€
    12% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Distinctive, broad, hatchet-shaped blade.
  • Broad, hatchet-shaped.
  • Used within podiatry.
  • Fitment No. 3.
  • Fits handles 3, 3L, 3 graduated, 5B, 7, 9, B3, and B3L.
Specifications
Blade Material Carbon Steel
Sterility Sterile
Quantity 100/Box
Packaging Type Individually Packaged