missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Residuals, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, Residuals, Ø38x15mm
Specifications
Specifications
| Chemical Name or Material | Arsenic/Phosphorus/Cobalt/Vanadium/Molybdenum/Titanium/Tungsten/Tin/Chromium/Copper/Aluminium/Nickel/Silicon/Carbon/Manganese |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Arsenic 0.01 %/Phosphorus 0.012 %/Cobalt 0.019 %/Vanadium 0.025 %/Molybdenum 0.075 %/Titanium 0.09 %/Tungsten 0.096 %/Tin 0.115 %/Chromium 0.127 %/Copper 0.15 %/Aluminium 0.155 %/Nickel 0.21 %/Silicon 0.25 %/Carbon 0.315 %/Manganese 0.59 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction