missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Residuals, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, Residuals, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Boron/Vanadium/Cobalt/Arsenic/Nitrogen/Zinc/Nickel/Phosphorus/Copper/Molybdenum/Aluminium/Chromium/Silicon/Carbon/Manganese |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0014 %/Vanadium 0.0033 %/Cobalt 0.0047 %/Arsenic 0.006 %/Nitrogen 0.0073 %/Zinc 0.012 %/Nickel 0.0197 %/Phosphorus 0.0205 %/Copper 0.0342 %/Molybdenum 0.0345 %/Aluminium 0.055 %/Chromium 0.07 %/Silicon 0.238 %/Carbon 0.364 %/Manganese 0.636 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction