missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Residuals, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, Residuals, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Vanadium/Phosphorus/Cobalt/Tin/Arsenic/Titanium/Molybdenum/Carbon/Nickel/Copper/Tungsten/Aluminium/Chromium/Silicon/Manganese |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Vanadium 0.012 %/Phosphorus 0.03 %/Cobalt 0.032 %/Tin 0.035 %/Arsenic 0.05 %/Titanium 0.075 %/Molybdenum 0.15 %/Carbon 0.16 %/Nickel 0.16 %/Copper 0.2 %/Tungsten 0.25 %/Aluminium 0.29 %/Chromium 0.34 %/Silicon 0.45 %/Manganese 0.76 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction