missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A1/CPA1 Antibody (rCPA1/8518), Novus Biologicals™
Mouse Monoclonal Antibody
249.00€ - 494.00€
Specifications
| Antigen | Carboxypeptidase A1/CPA1 |
|---|---|
| Clone | rCPA1/8518 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18644344
|
Novus Biologicals
NBP3-20336-100ug |
100 μg |
494.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670344
|
Novus Biologicals
NBP3-20336-20ug |
20 μg |
249.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carboxypeptidase A1/CPA1 Monoclonal antibody specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| Carboxypeptidase A1/CPA1 | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Mast Cell Markers | |
| 10mM PBS with 0.05% BSA | |
| 1357 | |
| IgG1 κ | |
| Protein A or G purified |
| rCPA1/8518 | |
| Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1 | |
| Recombinant full-length human Carboxypeptidase A1/CPA1 protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title