missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cardamonin
GREENER_CHOICE

Product Code. 18034812
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18034812 10 mg 10mg
18074042 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18034812 Supplier Tocris Bioscience™ Supplier No. 2509/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibitor of NF-κB activation; anti-inflammatory

Chalone analog that display anti-inflammatory activity. Inhibits NO and PGE2 production from LPS- and IFN-γ-induced RAW cells and inhibits TXB2 production via the COX-1 and COX-2 pathways. Inhibits NF-κB activation via inhibition of IκBα degradation and phosphorylation, IκB kinase activation and NF-κB nuclear translocation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19309-14-9
Molecular Formula C16H14O4

Specifications

Quantity 10 mg
Formula Weight 270.28
Purity >98%
Molecular Weight (g/mol) 270.284
Chemical Name or Material Cardamonin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.