Learn More
Carl Zeiss™ Axiovert™ Microscope Dust Cover
Product Code. 10661760
Change view
Click to view available options
For Use With (Equipment):
Axiovert 40/25 Microscope
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10661760 Axiovert 40/25 Microscope Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10661760 Supplier Carl Zeiss™ Supplier No. 4593150000000
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
L x W x H: 52 x 24 x 47cm
Specifications
Type Dust Cover
Dimensions (L x W x H) 20.47 x 9.44 x 18.5 in. (52 x 24 x 47 cm)
For Use With (Equipment) Axiovert 40/25 Microscope
Height (English) 18.5 in.
Height (Metric) 47 cm
Length (English) 20.47 in.
Length (Metric) 52 cm
Width (English) 9.44 in.
Width (Metric) 24 cm