missing translation for 'onlineSavingsMsg'
Learn More

Carphedone, TRC

Product Code. 17981745
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
Unit Size:
100mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17981745 100 mg 100mg
17963885 10 mg 10mg
17913245 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17981745 Supplier Toronto Research Chemicals Supplier No. TRCC184315100MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77472-70-9
Molecular Formula C12 H14 N2 O2
Molecular Weight (g/mol) 218.25
Synonym 1-Pyrrolidineacetamide, 2-oxo-4-phenyl-, 2-Oxo-4-phenyl-1-pyrrolidineacetamide, 2-(2-Oxo-4-phenylpyrrolidin-1-yl)acetamide, 4-Phenyl-2-oxopyrrolidine-1-acetamide, 4-Phenylpiracetam, Carphedon, Carphedone, Fonturacetam, Phenotropyl, Phenylpiracetam
IUPAC Name 2-(2-oxo-4-phenylpyrrolidin-1-yl)acetamide
SMILES NC(=O)CN1CC(CC1=O)c2ccccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H14N2O2/c13-11(15)8-14-7-10(6-12(14)16)9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H2,13,15)
Formula Weight 218.1055
Percent Purity >95%
Chemical Name or Material 4-Phenylpiracetam
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.