missing translation for 'onlineSavingsMsg'
Learn More

Carteolol Hydrochloride, TRC

Product Code. 17970385
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970385 100 mg 100mg
17900585 5 mg 5mg
17953515 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970385 Supplier Toronto Research Chemicals Supplier No. TRCC184450100MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51781-21-6
Molecular Formula C16 H24 N2 O3 . Cl H
Molecular Weight (g/mol) 328.83
Synonym 5-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-2(1H)-quinolinone Hydrochloride, (+/-)-Carteolol Hydrochloride, dl-Carteolol Hydrochloride;
IUPAC Name 5-[3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-1H-quinolin-2-one;hydrochloride
SMILES Cl.CC(C)(C)NCC(O)COc1cccc2NC(=O)CCc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H24N2O3.ClH/c1-16(2,3)17-9-11(19)10-21-14-6-4-5-13-12(14)7-8-15(20)18-13;/h4-6,11,17,19H,7-10H2,1-3H3,(H,18,20);1H
Formula Weight 328.1554
Percent Purity >95%
Chemical Name or Material Carteolol Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.