Learn More
Thermo Scientific™ Case for FH40TG Teleprobe
Product Code. 11930747 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
FH40TG Teleprobe
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11930747 FH40TG Teleprobe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11930747 Supplier Thermo Scientific™ Supplier No. 425405120
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Case for FH40TG Teleprobe without content.
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 5.9 in.
Material Aluminum
Width (English) 6.3 in.
Product Type Case
For Use With (Equipment) FH40TG Teleprobe
Dimensions 90 x 16 x 15 cm
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.