missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1 alpha Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-50030SS
This item is not returnable.
View return policy
Description
Casein Kinase 1 alpha Polyclonal antibody specifically detects Casein Kinase 1 alpha in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Casein Kinase 1 alpha | |
| Polyclonal | |
| Western Blot 1:10000, Immunohistochemistry 1:5000, Immunocytochemistry/ Immunofluorescence 1:500 - 1:1000 | |
| P48729 | |
| Chicken | |
| IgY purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS | |
| casein kinase 1, alpha 1, casein kinase I isoform alpha, CK1CKI-alpha, down-regulated in lung cancer, EC 2.7.11.1, HLCDGP1, PRO2975 | |
| Recombinant full length Casein Kinase 1 alpha (shortest isoform). [UniProt# P48729] | |
| 0.025 mL | |
| Protein Kinase, Wnt Signaling Pathway | |
| 1452 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction