missing translation for 'onlineSavingsMsg'
Learn More

Caspase-1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-76605-0.025mg

 View more versions of this product

Product Code. 18545873

  • 195.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Caspase-1 Polyclonal specifically detects Caspase-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Caspase-1
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
CASP-1, caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, caspase 1, apoptosis-related cysteine protease (interleukin 1, beta, caspase-1, EC 3.4.22, EC 3.4.22.36, ICEP45, IL-1 beta-converting enzyme, IL-1BC, IL1BC CASP1 nirs variant 1, IL1BCE, IL1B-convertase, interleukin 1-B converting enzyme, interleukin 1-beta convertase, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45
Rabbit
Affinity Purified
RUO
Primary
Depending on cell lines or tissues used, other cleavage products may be observed.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Western Blot
1 mg/mL
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL
NP_150634
CASP1
Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human Caspase-1. The immunogen is located within the last 50 amino acids of Caspase-1. Amino Acid Squence: SFEQPDGRAQMPTTER
0.025 mg
Apoptosis, Cancer, Caspases
834
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only