missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-3 Antibody (31A1067) - (Pro and Active) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 155 publications
305.00€ - 572.00€
Specifications
| Dilution | Western Blot 1 - 5 ug/ml, Simple Western 1:50, Flow Cytometry reported in scientific literature (PMID 27429862), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23840553), Immunohistochemistry-Paraffin 1:10 - 1:500. Use reported in scientific literature (PMID 28500555), Immunohistochemistry-Frozen 1:10 - 1:500. Use reported by customer review, Immunoblotting reported in scientific literature (PMID 28500555), Hematoxylin and Eosin Stain reported in scientific literature (PMID 28186963), Electron Microscopy reported in scientific literature (PMID 27450722), Immunohistochemistry Free-Floating reported in scientific literature (PMID 31771656), CyTOF-ready, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 32867814) |
|---|---|
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) |
| Classification | Monoclonal |
| Form | Purified |
| Target Species | Human, Mouse, Rat, Pig |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18766123
|
Novus Biologicals
NB100-56708SS |
305.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18767293
|
Novus Biologicals
NB100-56708 |
572.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse, Rat, Porcine, Chicken, Chinese Hamster, Mammal samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Hematoxylin & Eosin Stain, Electron Microscopy, Immunohistochemistry Free-Floating, CyTOF-ready, Knockout Validated, Knockdown Validated.Specifications
| Western Blot 1 - 5 ug/ml, Simple Western 1:50, Flow Cytometry reported in scientific literature (PMID 27429862), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23840553), Immunohistochemistry-Paraffin 1:10 - 1:500. Use reported in scientific literature (PMID 28500555), Immunohistochemistry-Frozen 1:10 - 1:500. Use reported by customer review, Immunoblotting reported in scientific literature (PMID 28500555), Hematoxylin and Eosin Stain reported in scientific literature (PMID 28186963), Electron Microscopy reported in scientific literature (PMID 27450722), Immunohistochemistry Free-Floating reported in scientific literature (PMID 31771656), CyTOF-ready, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 32867814) | |
| Monoclonal | |
| Human, Mouse, Rat, Pig | |
| Protein G purified |
| Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) | |
| Purified | |
| CASP3 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title