Learn More
Caspase 4/p20/p10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11856-1-AP-20UL
This item is not returnable.
View return policy
Description
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.Specifications
| Caspase 4/p20/p10 | |
| Polyclonal | |
| Unconjugated | |
| CASP4 | |
| CASP 4, CASP4, Caspase 4, Caspase4, ICE(rel) II, ICE(rel)II, ICEREL II, ICH 2, ICH2, Mih1/TX, Protease ICH 2, Protease TX, TX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 837 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49662 | |
| CASP4 | |
| Caspase 4/p20/p10 Fusion Protein Ag2429 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |