Learn More
R&L Enterprises™ Tripod Shape Retort Stand Base
Product Code. 12338139 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Holds:
Rod Centre: 135mm
Rod Centre: 175mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
12338139 Rod Centre: 135mm Each
12916806 Rod Centre: 175mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12338139
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Excellent option for supporting retort stands in the laboratory. R&L Enterprises™ Tripod Shape Retort Stand Base are made from cast iron.
  • Painted blue
  • With rubber feet
  • Central rod hole is tapped
  • Without rod
Specifications
Material Cast Iron
Holds Rod Centre: 135mm
Includes With Rubber Feet
Product Type Tripod Spread Base
Color Blue