Learn More
Catalase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21260-1-AP-20UL
 View more versions of this product
Product Code. 16850875
  • 145.00€ / 20µL
Description
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kD in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara′s disease.
Specifications
Specifications
Catalase
Polyclonal
Unconjugated
CAT
CAT, catalase
Rabbit
Antigen Affinity Chromatography
RUO
12359, 24248, 847
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04040, P04762, P24270
CAT
Catalase Fusion Protein Ag15383
20 μL
Primary
Rat, Mouse, Human