missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Catalase Recombinant Rabbit Monoclonal Antibody (24GB4295)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557355
This item is not returnable.
View return policy
Description
Catalase Recombinant Monoclonal Antibody for Western Blot, Flow
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.
Specifications
| Catalase | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04040 | |
| Cat | |
| A synthesized peptide derived from human Catalase (aa 425-438). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4295 | |
| Unconjugated | |
| Cat | |
| 2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 847 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur