Learn More
Löser™ Freezing Needle for Osmometer
Product Code. 11897714
Change view
Click to view available options
For Use With (Equipment):
All Osmometers 50 to 200 μL Sample Volume
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11897714 All Osmometers 50 to 200 μL Sample Volume Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11897714 Supplier Löser™ Supplier No. 1.05.0400
  • ONLINE EXCLUSIVE PRICE
    10.35€
    5.46€ / Pack of 1
    Save 4.89€
    47% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Provides initial freezing point trigger for osmometers. Freezing Needling for Osmometer can be used with all osmometers with a sample volume between 50 and 200μL.
  • Needle for freezing initiation of all Osmometers with sample volume 50 to 200μL.
Specifications
Product Type Freezing Needle
For Use With (Equipment) All Osmometers 50 to 200 μL Sample Volume
For Use With (Application) Triggering the freezing with freezing point Osmometers