missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin L/V/K/H Antibody (SR1633), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21720-100ul
This item is not returnable.
View return policy
Description
Cathepsin L/V/K/H Monoclonal antibody specifically detects Cathepsin L/V/K/H in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Specifications
| Cathepsin L/V/K/H | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| SR1633 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| ACC4, ACC-4, ACC5, ACC-5, aleurain, cathepsin B3, cathepsin BA, cathepsin K, cathepsin L1, cathepsin L2, cathepsin L2, preproprotein, Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin U, Cathepsin X, CATL, CATL2, CPSB, CTS02, CTSL1, CTSL2, CTSO, CTSO1, CTSO2, CTSU, Major excreted protein, MEP, N-benzoylarginine-beta-naphthylamide hydrolase, PKND, procathepsin L, PYCD | |
| A synthesized peptide derived from human Cathepsin L/V/K/H (Uniprot #: P07711, O60911, P43235, P09668) | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 1514 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction