missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin O Antibody (MM0131-9G31), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12083APC
This item is not returnable.
View return policy
Description
Cathepsin O Monoclonal antibody specifically detects Cathepsin O in Human samples. It is validated for Western Blot
Specifications
| Cathepsin O | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1519 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| MM0131-9G31 | |
| Western Blot | |
| cathepsin O, CTSO1, EC 3.4.22.42 | |
| Human recombinant Cathepsin O | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction