missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin S Antibody (1F9), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42634DL594
This item is not returnable.
View return policy
Description
Cathepsin S Monoclonal antibody specifically detects Cathepsin S in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Cathepsin S | |
| Monoclonal | |
| DyLight 594 | |
| cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886 | |
| Recombinant human Cathepsin S (17-331aa) purified from E. coli | |
| 0.1 mL | |
| Immunology | |
| 1520 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Immunofluorescence | |
| 1F9 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction