missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-2 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99448UV
This item is not returnable.
View return policy
Description
Caveolin-2 Polyclonal antibody specifically detects Caveolin-2 in Human samples. It is validated for Western Blot
Specifications
| Caveolin-2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| CAV, caveolae protein, 20-kD, caveolin 2, caveolin 2 isoform a and b, caveolin-2, MGC12294 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Caveolin-2. | |
| 0.1 mL | |
| Caveolae Markers, Membrane Trafficking and Chaperones, Neuroscience | |
| 858 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction