Learn More
Karl Hecht™ Cavity Slide/Plates
Product Code. 11740555
Change view
Click to view available options
Description: 16mm cavity diameter, 1.6mm cavity depth, polished cavities, Dimensions: 140mm x 85mm, Surface Treatment: Frosted, Thickness: 3mm, Well Count: 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11740555 Description: 16mm cavity diameter, 1.6mm cavity depth, polished cavities, Dimensions: 140mm x 85mm, Surface Treatment: Frosted, Thickness: 3mm, Well Count: 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11740555 Supplier Karl Hecht™ Supplier No. 42418010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Perfectly suited for holding liquids and cultures for microscopic examination. Karl Hecht™ Cavity Slides/Plates are available with one or more cavities and in single slide or plate format.
  • Microscope slides/plates with one or more cavities.
  • Ground edges.
Specifications
Length (Metric) 140 mm
Width (Metric) 85 mm
Frosted Yes
Thickness (Metric) 3 mm
Type Microscope Slide
Well Count 24
Description 16mm cavity diameter, 1.6mm cavity depth, polished cavities
Dimensions (L x W) 140 x 85 mm