missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92934-0.02ml
This item is not returnable.
View return policy
Description
CBX4 Polyclonal antibody specifically detects CBX4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| CBX4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50-1:200 | |
| chromobox homolog 4, chromobox homolog 4 (Drosophila Pc class), Chromobox protein homolog 4, Drosophila), E3 SUMO-protein ligase CBX4, hPC2, NS5ATP1-binding protein 16, Pc class 2 homolog, PC2, Polycomb 2 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 401-560 of human CBX4 (NP_003646.2). AVGLNLSHVRKRCLSETHGEREPCKKRLTARSISTPTCLGGSPAAERPADLPPAAALPQPEVILLDSDLDEPIDLRCVKTRSEAGEPPSSLQVKPETPASAAVAVAAAAAPTTTAEKPPAEAQDEPAESLSEFKPFFGNIIITDVTANCLTVTFKEYVTV | |
| 0.02 mL | |
| Apoptosis | |
| 8535 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction