missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCAR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22638-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DBC1 is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5′ CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.Spécification
| CCAR2 | |
| Polyclonal | |
| Unconjugated | |
| CCAR2 | |
| ccar2, DBC.1, DBC-1, Deleted in breast cancer gene 1 protein, KIAA1967, NET35, p30 DBC, Protein KIAA1967 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 219158, 306007, 57805 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N163, Q8VDP4 | |
| CCAR2 | |
| CCAR2 Fusion Protein Ag18374 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu