missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC102B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-87139
This item is not returnable.
View return policy
Description
CCDC102B Polyclonal specifically detects CCDC102B in Human samples. It is validated for Western Blot.
Specifications
| CCDC102B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79839 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| ACY1L, aminoacylase 1-like, AN, C18orf14, chromosome 18 open reading frame 14, coiled-coil domain containing 102B, coiled-coil domain-containing protein 102B, DKFZp434K1426, DKFZp686I08254, FLJ23594, HsT1731, MGC161726, MGC161728 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of CCDC102B | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction