missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC109A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12716-100ul
This item is not returnable.
View return policy
Description
CCDC109A Polyclonal antibody specifically detects CCDC109A in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| CCDC109A | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate | |
| C10orf42, coiled-coil domain containing 109A, coiled-coil domain-containing protein 109A, FLJ46135 | |
| Produced in rabbits immunized with E. coli-derived Human CCDC109A fragment | |
| 100 μg | |
| Cardiovascular Biology, Endocrinology | |
| 90550 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction