missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584460
This item is not returnable.
View return policy
Description
Immunogen sequence: KPEPVIEEVD LANLAPRKPD WDLKRDVAKK LEKLKKRTQR AIAELIRERL KGQEDSLASA VDAATEQKTC DS Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 93%.
The exact function of this protein remains unknown.
Specifications
| CCDC12 | |
| Polyclonal | |
| Unconjugated | |
| CCDC12 | |
| 2700094L05Rik; C76605; CCDC12; coiled-coil domain containing 12; coiled-coil domain-containing protein 12; RGD1306081 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 151903 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WUD4 | |
| CCDC12 | |
| Recombinant protein corresponding to Human CCDC12. Recombinant protein control fragment (Product #RP-107016). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction