missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC28A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563069
This item is not returnable.
View return policy
Description
Immunogen sequence: LARLNLELYG ELEELPEDKR KTASDSNLDR LLSDLEELNS SIQKLHLADA QDVPNTSA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 88%.
This gene encodes a coiled-coil domain containing protein. Although the specific function of this gene has not yet been determined, this gene is a known translocation partner of nucleoporin 98 in acute leukemias. The resulting fusion gene produces a nucleoporin 98-coiled-coil domain-containing protein 28A chimeric protein which may be involved in promoting myeloproliferative neoplasms.
Specifications
| CCDC28A | |
| Polyclonal | |
| Unconjugated | |
| CCDC28A | |
| 1700009P13Rik; AI480677; C6orf80; CCDC28A; CCRL1AP; chemokine C-C motif receptor-like 1 adjacent; coiled-coil domain containing 28A; coiled-coil domain-containing protein 28A; RGD1310326 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25901 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IWP9 | |
| CCDC28A | |
| Recombinant protein corresponding to Human CCDC28A. Recombinant protein control fragment (Product #RP-101716). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction