missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC28B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11530-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene localizes to centrosomes and basal bodies. It interacts and colocalizes with several proteins associated with Bardet-Biedl syndrome (BBS).Specifications
| CCDC28B | |
| Polyclonal | |
| Unconjugated | |
| CCDC28B | |
| CCDC28B, RP4 622L5.5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66264, 682445, 79140 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CEG5, Q9BUN5 | |
| Ccdc28b | |
| CCDC28B Fusion Protein Ag2105 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction