missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC50 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | CCDC50 |
|---|---|
| Clone | 2F12G7 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235042
|
Proteintech
68369-1-IG-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237832
|
Proteintech
68369-1-IG-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CCDC50 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCDC50. This antibody recognizes human antigen. The CCDC50 antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.Specifications
| CCDC50 | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 152137 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2F12G7 | |
| Western Blot, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| C3orf6; DFNA44; YMER | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title