Learn More
CCDC93 Monoclonal Antibody (OTI1A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800470S
Description
CCDC93 Monoclonal antibody specifically detects CCDC93 in Canine, Human samples. It is validated for Western BlotSpecifications
| CCDC93 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q567U6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 476119, 54520 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1A10 | |
| Unconjugated | |
| CCDC93 | |
| 4633402D15Rik; 9230102M16Rik; CCDC93; coiled-coil domain containing 93; coiled-coil domain-containing protein 93 | |
| Human recombinant protein fragment corresponding to amino acids 293-631 of human CCDC93 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.