missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCL14 Recombinant Rabbit Monoclonal Antibody (015), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
256.00€ - 512.00€
Specifications
| Antigen | CCL14 |
|---|---|
| Clone | 015 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17769813
|
Invitrogen™
MA543468 |
50 μL |
256.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17799613
![]() |
Invitrogen™
MA543065 |
100 μL |
512.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specificity: Human CCL14
This gene, CCL14, is one of several CC cytokine genes clustered on 17q11.2. The CC cytokines are secreted proteins characterized by two adjacent cysteines. The cytokine encoded by this gene induces changes in intracellular calcium concentration and enzyme release in monocytes. This gene expresses both monocistronic and bicistronic transcripts. Bicistronic transcripts include the upstream cytokine gene CCL15. In addition, CCL14 undergoes alternative splicing in the bicistronic transcripts; it is unknown if its monocistronic transcript is also subject to alternative splicing.Specifications
| CCL14 | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| C-C motif chemokine 14; C-C motif chemokine ligand 14; CC-1; CC-3; CCL14; chemokine (C-C motif) ligand 14; chemokine CC-1/CC-3; chemokine CC-3; CKB1; H HCC-1(1-74); HCC-1; HCC-1(1-74); HCC-1(3-74); HCC-1(4-74); HCC-1(9-74); HCC-1/HCC-3; HCC-3; hemofiltrate CC chemokine 1; MCIF; NCC2; NCC-2; new CC chemokine 2; SCYA14; SCYL2; small inducible cytokine subfamily A (Cys-Cys), member 14; small-inducible cytokine A14; SY14 | |
| CCL14 | |
| IgG | |
| Protein A | |
| Antibody |
| 015 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q16627 | |
| 6358 | |
| Recombinant Human CCL14 Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| CCL14 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title